missing translation for 'onlineSavingsMsg'
Learn More

(2R)-(-)-Glycidyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T16115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16115G 5 g
T161125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16115G Supplier TCI America Supplier No. T16115G

Chemical Identifiers

CAS 113826-06-5
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00010834
InChI Key NOQXXYIGRPAZJC-SECBINFHSA-N
Synonym 2r---glycidyl tosylate, r-glycidyl tosylate, r---glycidyltosylate, --glycidyl tosylate, 2r---glycidyltosylate, r---glycidyl tosylate, r-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6399, 2r-glycidyl tosylate, 2r-oxiran-2-ylmethyl 4-methylbenzenesulfonate
PubChem CID 154187
IUPAC Name [(2R)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2

Specifications

Melting Point 48°C
Color White
Quantity 5 g
Formula Weight 228.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2R)-(-)-Glycidyl p-Toluenesulfonate
RTECSNumber RR0510000
TSCA No