missing translation for 'onlineSavingsMsg'
Learn More

(2R,5R)-2,5-Hexanediol 98.0+%, TCI America™

Catalog No. H10321G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10321G Supplier TCI America Supplier No. H10321G
Only null left

Chemical Identifiers

CAS 17299-07-9
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00142339
InChI Key OHMBHFSEKCCCBW-PHDIDXHHSA-N
Synonym 2r,5r-2,5-hexanediol, 2r,5r-hexane-2,5-diol, 2,5-hexanediol, 2r,5r, 2r,5r---2,5-hexanediol, hx2, 2r,5r-hexanediol, 2r,5r-2,5-dihydroxyhexane, 2r,5r-2,5-hexanediol sum of enantiomers, gc
PubChem CID 2733360
IUPAC Name (2R,5R)-hexane-2,5-diol
SMILES CC(CCC(C)O)O

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2R,5R)-2,5-Hexanediol
EINECSNumber (2)-0240
TSCA No