missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 395010050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC395010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

98760-08-8
263.34
NVPOUMXZERMIJK-UHFFFAOYNA-N
9813904
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
C15H21NO3
MFCD00671705,MFCD02258997
2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate

Specifications

(2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
97.5
97.5% min. (HPLC)
C15H21NO3
5g
Solubility in water: <0.1%
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
263.34
263.34
98760-08-8
100.0
Glass bottle
MFCD00671705,MFCD02258997
2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
NVPOUMXZERMIJK-UHFFFAOYNA-N
tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate
9813904
98%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions