missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%

Catalog No. AAH52806MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52806MD 250 mg
AAH5280603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52806MD Supplier Thermo Scientific Chemicals Supplier No. H52806MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73397-28-1 | C12H23NO5 | 261.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73397-28-1
Molecular Formula C12H23NO5
Molecular Weight (g/mol) 261.318
MDL Number MFCD04972275
InChI Key DJZCWTDKDFJARG-DTWKUNHWSA-N
Synonym boc-2r,3s-3-amino-2-hydroxy-5-methylhexanoicacid, 2r,3s-3-boc-amino-2-hydroxy-5-methylhexanoic acid, n-boc-2r,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2r,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, 2r,3s-2-hydroxy-3-tert-butoxycarbonylamino-5-methylhexanoic acid
PubChem CID 10377964
IUPAC Name (2R,3S)-2-hydroxy-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)CC(C(C(=O)O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 261.32
Percent Purity 97%
Chemical Name or Material (2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only