missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-Tartranilic Acid 98.0+%, TCI America™

Catalog No. T17025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17025G 5 g
T17021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17025G Supplier TCI America Supplier No. T17025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3019-58-7
Molecular Formula C10H11NO5
Molecular Weight (g/mol) 225.20
MDL Number MFCD01321193
InChI Key ZWXNRJCDXZFNLJ-UHFFFAOYNA-N
Synonym (2R,3R)-2,3-Dihydroxy-3-(phenylcarbamoyl)propionic Acid
PubChem CID 11407500
IUPAC Name 2,3-dihydroxy-3-(phenylcarbamoyl)propanoic acid
SMILES OC(C(O)C(=O)NC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 225.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2R,3R)-Tartranilic Acid [for optical resolution]