missing translation for 'onlineSavingsMsg'
Learn More

2H-1,4-Benzoxazin-3(4H)-one 98.0+%, TCI America™

Catalog No. B45535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45535G 5 g
B455325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45535G Supplier TCI America Supplier No. B45535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5466-88-6
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00158536
InChI Key QRCGFTXRXYMJOS-UHFFFAOYSA-N
Synonym 2h-1,4-benzoxazin-3 4h-one, 2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3-one, 3-oxo-4h-benzo 1,4 oxazine, 3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3-ol, 2h-1,4-benzoxazin-3 4h-one 8ci 9ci, 4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazine-3 4h-one, 2h,4h-benzo e 1,4-oxazaperhydroin-3-one
PubChem CID 72757
IUPAC Name 4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=CC=CC=C2O1

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 149.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2H-1,4-Benzoxazin-3(4H)-one
RTECSNumber DM3270000
TSCA No