Learn More
Celltreat Scientific Products 25mm Round Cover Glass, Sterile
Supplier: Celltreat Scientific Products 229174
CELLTREAT Scientific Products 25mm Round Cover Glass. High-quality, optically clear borosilicate glass cover slips are ideal for specialty microscopy and imaging. Ideal for use with dishes and multiple well plates and anywhere a square cover glass is unsuitable. Consistent flatness and scratch-free. Resistant to organic solvents. 25mm Round Cover Glass can be used with a 6 well glass bottom plate (Part No. 229107). Cover Glass is #1.5 thickness. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged in a hinged plastic storage case, sterile, 100/case