Learn More
Celltreat Scientific Products 25mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile pipette
Numéro de catalogue. 50969837
Encompass
CELLTREAT Scientific Products 25mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Negative graduations of -10mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic wrappers. 50/bag, 200/case.
Numéro de catalogue. 50969837 Fournisseur Celltreat Scientific Products Code fournisseur 229025B
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
CELLTREAT Scientific Products 25mL Serological Pipet. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Negative graduations of -10mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic wrappers. 50/bag, 200/case.