Learn More
Alkali Scientific 20 Liter Bead Waterbath, Includes beads, Lab Armor
Catalog No. 502336095
Encompass
Our Eco-Friendly Lab Armor Bead Bath with Beads included control the temperature of samples in metal beads instead of water or other liquid. This avoids possible contamination from water or other liquid warming mediums while allowing for easier sample organization without the need for racks or clamps. Bead baths are also easier to maintain as they do not need to be cleaned. While bath beads can be used in most stationary water baths, these baths are specifically designed for use with beads and provide improved temperature uniformity. They are indispensable in applications where contamination or sterility is a concern.
Catalog No. 502336095 Supplier Alkali Scientific Supplier No. WB713
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Our Eco-Friendly Lab Armor Bead Bath with Beads included control the temperature of samples in metal beads instead of water or other liquid. This avoids possible contamination from water or other liquid warming mediums while allowing for easier sample organization without the need for racks or clamps. Bead baths are also easier to maintain as they do not need to be cleaned. While bath beads can be used in most stationary water baths, these baths are specifically designed for use with beads and provide improved temperature uniformity. They are indispensable in applications where contamination or sterility is a concern.