Chemglass Inc Reaction Vessel Lid, 4-Neck, 45/50 Center Neck, 3-24/40 Side Necks, 200mm Schott Flange
Numéro de catalogue. CG194107
Reaction Vessel Lid, 4-Neck, 45/50 Center Neck, 3-24/40 Side Necks, 200mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.
Numéro de catalogue.
CG194107
Fournisseur
Chemglass IncCode fournisseur
CG194107
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Reaction Vessel Lid, 4-Neck, 45/50 Center Neck, 3-24/40 Side Necks, 200mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.