missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone 98.0+%, TCI America™

Catalog No. U0006500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0006500ML 500 mL
U000625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0006500ML Supplier TCI America Supplier No. U0006500ML

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 500 mL
Formula Weight 170.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Undecanone
EINECSNumber (2)-0542
RTECSNumber YQ2820000
TSCA Yes