missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate 98.0+%, TCI America™

Catalog No. T25445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25445G 5 g
T25441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25445G Supplier TCI America Supplier No. T25445G
Only null left

Chemical Identifiers

CAS 1001067-09-9
Molecular Formula C8H14N4O4Si
Molecular Weight (g/mol) 258.31
MDL Number MFCD11042686
InChI Key BHTJYIDOVYPNKR-UHFFFAOYSA-N
Synonym 3-Nitro-1H-1,2,4-triazole-1-carboxylic Acid 2-(Trimethylsilyl)ethyl Ester, 3-Nitro-1-[2-(trimethylsilyl)ethoxycarbonyl]-1H-1,2,4-triazole, Teoc-NT
PubChem CID 23731110
IUPAC Name 2-(trimethylsilyl)ethyl 3-nitro-1H-1,2,4-triazole-1-carboxylate
SMILES C[Si](C)(C)CCOC(=O)N1C=NC(=N1)[N+]([O-])=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 258.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate
TSCA No
Recommended Storage Freezer