missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethanol 96.0+%, TCI America™

Catalog No. T14415ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14415ML 5 mL
T144125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14415ML Supplier TCI America Supplier No. T14415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2916-68-9
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.251
MDL Number MFCD00002825
InChI Key ZNGINKJHQQQORD-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
PubChem CID 18013
IUPAC Name 2-trimethylsilylethanol
SMILES C[Si](C)(C)CCO

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 mL
UN Number 1993
Formula Weight 118.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)ethanol
RTECSNumber KM5480000
TSCA No