missing translation for 'onlineSavingsMsg'
Learn More

2-Trimethylsilyl-1,3-dithiane 96.0+%, TCI America™

Catalog No. T15145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15145G 5 g
T151425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15145G Supplier TCI America Supplier No. T15145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13411-42-2
Molecular Formula C7H16S2Si
Molecular Weight (g/mol) 192.41
MDL Number MFCD00006655
InChI Key BTTUMVHWIAXYPJ-UHFFFAOYSA-N
Synonym 2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane
PubChem CID 83413
IUPAC Name (1,3-dithian-2-yl)trimethylsilane
SMILES C[Si](C)(C)C1SCCCS1

Specifications

Color Yellow
Boiling Point 77°C
Quantity 5 g
Formula Weight 192.41
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Trimethylsilyl-1,3-dithiane