Learn More
2-(Trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals
Description
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.
Solubility
Soluble in most organic solvents.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.28 |
| Boiling Point | 139°C to 141°C |
| Flash Point | 42°C (108°F) |
| Refractive Index | 1.418 |
| Quantity | 1 g |
| UN Number | UN1993 |
| Beilstein | 3112 |
| Solubility Information | Soluble in most organic solvents. |
| Formula Weight | 147.1 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only