missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%

Catalog No. AAH6405403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6405403 1 g
AAH6405406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6405403 Supplier Thermo Scientific Chemicals Supplier No. H6405403

CAS: 386704-12-7 | C7H4F3NO | 175.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 386704-12-7
Molecular Formula C7H4F3NO
Molecular Weight (g/mol) 175.11
MDL Number MFCD01862647
InChI Key MRPAGRCGPAXOGS-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
PubChem CID 2777762
IUPAC Name 6-(trifluoromethyl)pyridine-3-carbaldehyde
SMILES FC(F)(F)C1=NC=C(C=O)C=C1

Specifications

Melting Point 52°C to 56°C
Flash Point 93°C (199°F)
Quantity 1 g
UN Number UN2811
Sensitivity Air sensitive
Formula Weight 175.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-carboxaldehyde
Hazard Category H301-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only