missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)isonicotinic Acid 98.0+%, TCI America™

Catalog No. T3269200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3269200MG 200 mg
T32691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3269200MG Supplier TCI America Supplier No. T3269200MG

Chemical Identifiers

CAS 131747-41-6
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07774132
InChI Key BZFGKBQHQJVAHS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl
PubChem CID 14761447
IUPAC Name 2-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1)C(F)(F)F

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 200 mg
Formula Weight 191.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)isonicotinic Acid