missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America T13115G
Specifications
| 2-(Trifluoromethyl)benzyl Alcohol | |
| Yellow | |
| C8H7F3O | |
| 5 g | |
| TWQNSHZTQSLJEE-UHFFFAOYSA-N | |
| [2-(trifluoromethyl)phenyl]methanol | |
| 67666 | |
| ≥98.0% (GC) |
| 346-06-5 | |
| 70°C | |
| MFCD00004621 | |
| 2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol | |
| OCC1=CC=CC=C1C(F)(F)F | |
| 176.14 | |
| 176.14 | |
| Liquid |
Chemical Identifiers
| 346-06-5 | |
| 176.14 | |
| TWQNSHZTQSLJEE-UHFFFAOYSA-N | |
| 67666 | |
| OCC1=CC=CC=C1C(F)(F)F |
| C8H7F3O | |
| MFCD00004621 | |
| 2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol | |
| [2-(trifluoromethyl)phenyl]methanol |
Safety and Handling
TSCA : No