missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)phenol 96.0+%, TCI America™

Catalog No. T21565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21565G 5 g
T215625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21565G Supplier TCI America Supplier No. T21565G

Chemical Identifiers

CAS 32858-93-8
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00236324
InChI Key GQWMNVOVQZIPJC-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
PubChem CID 2777299
IUPAC Name 2-(trifluoromethoxy)phenol
SMILES C1=CC=C(C(=C1)O)OC(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 178.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)phenol