missing translation for 'onlineSavingsMsg'
Learn More

2'-(Trifluoromethoxy)acetophenone 98.0+%, TCI America™

Catalog No. T27945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27945G 5 g
T279425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27945G Supplier TCI America Supplier No. T27945G

Chemical Identifiers

CAS 220227-93-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00236320
InChI Key XTXSTESGCXKUIH-UHFFFAOYSA-N
Synonym 2'-trifluoromethoxy acetophenone, 1-2-trifluoromethoxy phenyl ethanone, 1-2-trifluoromethoxy phenyl ethan-1-one, 1-acetyl-2-trifluoromethoxy benzene, pubchem4295, 2-trifluoromethoxy acetophenone98, 1-2-trifluoromethoxyphenyl ethanone, 2'-trifluoromethoxy acetophenone98
PubChem CID 2777176
IUPAC Name 1-[2-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=CC=C1OC(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-(Trifluoromethoxy)acetophenone