missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethylamine, 94%

Catalog No. AAA1170022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1170022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1170022 Supplier Thermo Scientific Chemicals Supplier No. A1170022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27757-85-3 | C5H7NS | 113.178 g/mol

2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thiophenemethylamine is Reactant involved in synthesis of: Triazole-linked-thiopene conjugates. It is used for biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein.It is a reactant involved in studies of organocatalyzed asymmetric reductive amination of ketones.

Solubility
Easily soluble in ethanol and Chloroform.

Notes
Air Sensitive. Store under dry inert gas. Store away from air.

Chemical Identifiers

CAS 27757-85-3
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005460
InChI Key FKKJJPMGAWGYPN-UHFFFAOYSA-N
Synonym 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
PubChem CID 34005
IUPAC Name thiophen-2-ylmethanamine
SMILES C1=CSC(=C1)CN

Specifications

Density 1.1
Boiling Point 85°C to 88°C (14 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.567
Quantity 100 g
UN Number UN2735
Beilstein 106296
Sensitivity Air Sensitive
Solubility Information Easily soluble in ethanol and Chloroform.
Formula Weight 113.19
Percent Purity 97%
Chemical Name or Material 2-Thiophenemethylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 248-639-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only