missing translation for 'onlineSavingsMsg'
Learn More

2-Thiopheneacetamide, 98%

Catalog No. AAL1922122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1922122 100 g
AAL1922106 5 g
AAL1922114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1922122 Supplier Thermo Scientific Chemicals Supplier No. L1922122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4461-29-4 | C6H7NOS | 141.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4461-29-4
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD00017556
InChI Key UUPZTFTUZUQRQT-UHFFFAOYSA-N
Synonym 2-thiophen-2-yl acetamide, thiophene-2-acetamide, 2-thiopheneacetamide, 2-2-thienyl acetamide, thiophen-2-acetamide, thiophen-2-yl-acetamide, 2-thienylacetamide, thiolacet-amide, 2-thienyl acetamide, acmc-1ajdb
PubChem CID 78208
IUPAC Name 2-thiophen-2-ylacetamide
SMILES C1=CSC(=C1)CC(=O)N

Specifications

Melting Point 145°C to 148°C
Quantity 100 g
Beilstein 114558
Formula Weight 141.19
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Thiopheneacetamide
Hazard Category H301
Hazard Statement GHS H Statement

H301

Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
EINECSNumber 224-714-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only