missing translation for 'onlineSavingsMsg'
Learn More

2-Tetralone, 99%

Numéro de catalogue. p-7043503
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0105306 5 g
AAL0105314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0105306 Fournisseur Thermo Scientific Chemicals Code fournisseur L0105306
Il en reste null

CAS: 530-93-8 | C10H10O | 146.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Spécifications

Melting Point 18°C
Density 1.106
Boiling Point 240°C
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 5 g
Beilstein 509386
Sensitivity Air sensitive
Formula Weight 146.19
Percent Purity 99%
Chemical Name or Material 2-Tetralone
Afficher plus de résultats Afficher moins de résultats
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-498-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only