missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-ethylphenol 97.0+%, TCI America™

Catalog No. B181025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B181025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B181025G Supplier TCI America Supplier No. B181025G
Only null left

Chemical Identifiers

CAS 96-70-8
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00069413
InChI Key LZHCVNIARUXHAL-UHFFFAOYSA-N
PubChem CID 7309
IUPAC Name 2-tert-butyl-4-ethylphenol
SMILES CCC1=CC=C(O)C(=C1)C(C)(C)C

Specifications

Color White-Yellow
Boiling Point 248°C
Quantity 25 g
Formula Weight 178.28
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-tert-Butyl-4-ethylphenol
EINECSNumber (3)-0521
TSCA Yes