missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 98.0+%, TCI America™
Supplier: TCI America B10895G
Specifications
| 2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine [Boc Agent for Peptides Synthesis] | |
| 51°C | |
| MFCD00006080 | |
| s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester | |
| CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1 | |
| 240.32 | |
| 240.32 | |
| Crystalline Powder |
| 41840-28-2 | |
| C11H16N2O2S | |
| 5 g | |
| POTDIELOEHTPJN-UHFFFAOYSA-N | |
| tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate | |
| 148428 | |
| ≥98.0% (T) |
Chemical Identifiers
| 41840-28-2 | |
| 240.32 | |
| POTDIELOEHTPJN-UHFFFAOYSA-N | |
| 148428 | |
| CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1 |
| C11H16N2O2S | |
| MFCD00006080 | |
| s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester | |
| tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate |
Safety and Handling
EINECSNumber : (5)-3821
TSCA : No
Recommended Storage : Refrigerator