missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine 98.0+%, TCI America™

Catalog No. p-7118845
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10895G 5 g
B108925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10895G Supplier TCI America Supplier No. B10895G
Only null left

Chemical Identifiers

CAS 41840-28-2
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD00006080
InChI Key POTDIELOEHTPJN-UHFFFAOYSA-N
Synonym s-boc-2-mercapto-4,6-dimethylpyrimidine, ccris 2600, 2-tert-butoxycarbonylthio-4,6-dimethylpyrimidine, 2-boc-thio-4,6-dimethylpyrimidine, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiocarbonate, tert-butyl s-4,6-dimethylpyrimidin-2-yl thiolcarbonate, o-tert-butyl s-4,6-dimethylpyrimidin-2-yl carbonothioate, carbonothioic acid, o-1,1-dimethylethyl s-4,6-dimethyl-2-pyrimidinyl ester, tert-butyl 4,6-dimethylpyrimidin-2-yl sulfanylformate, thiocarbonic acid, o-tert-butyl s-4,6-dimethyl-2-pyrimidinyl ester
PubChem CID 148428
IUPAC Name tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate
SMILES CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1

Specifications

Melting Point 51°C
Quantity 5 g
Formula Weight 240.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonylthio)-4,6-dimethylpyrimidine [Boc Agent for Peptides Synthesis]
EINECSNumber (5)-3821
TSCA No
Recommended Storage Refrigerator