accessibility menu, dialog, popup

UserName

CH3CH3NSOOCH3H3CCH3N

CAS RN 41840-28-2

IUPAC Name: tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate
Synonyms: tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate
Molecular Weight (g/mol): 240.32
Molecular Formula: C11H16N2O2S
InChi Key: POTDIELOEHTPJN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)