missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile 98.0+%, TCI America™

Catalog No. B09885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09885G 5 g
B098825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09885G Supplier TCI America Supplier No. B09885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58632-95-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.266
MDL Number MFCD00001863
InChI Key QQWYQAQQADNEIC-PTNGSMBKSA-N
Synonym 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
PubChem CID 5868400
IUPAC Name tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
SMILES CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1

Specifications

Melting Point 88°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 246.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile
EINECSNumber (3)-3524
TSCA Yes
Recommended Storage Refrigerator