missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. B4552200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4552200MG 200 mg
B45521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4552200MG Supplier TCI America Supplier No. B4552200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 302964-02-9
Molecular Formula C9H12N2O4S
Molecular Weight (g/mol) 244.27
MDL Number MFCD06796614
InChI Key QNFLEDLPOVONCN-UHFFFAOYSA-N
Synonym 2-(Boc-amino)thiazole-5-carboxylic Acid
PubChem CID 22271442
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-1,3-thiazole-5-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1=NC=C(S1)C(O)=O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 244.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic Acid