missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxaloyl chloride, 95%

Catalog No. AC163760010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163760010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC163760010 Supplier Thermo Scientific Chemicals Supplier No. 163760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54745-92-5 | C9H5ClN2O | 192.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54745-92-5
Molecular Formula C9H5ClN2O
Molecular Weight (g/mol) 192.6
InChI Key SOPDQKNXOCUBSR-UHFFFAOYSA-N
Synonym 2-quinoxalinecarbonyl chloride, 2-quinoxalinecarbonylchloride, 2-quinoxaloyl chloride, 2-quinoxaloylchloride, 2-quinoxalinylcarbonyl chloride, 2-quinoxaline carbonyl chloride, 2-quinoxalinecarbonyl chloride 7ci, 9ci
PubChem CID 2734681
IUPAC Name quinoxaline-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl

Specifications

Melting Point 113.0°C to 115.0°C
Color Colorless to Yellow
CAS Min % 94.0
Physical Form Liquid
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Formula Weight 192.6
Percent Purity 95%
Chemical Name or Material 2-Quinoxaloyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-315-9

RUO – Research Use Only