missing translation for 'onlineSavingsMsg'
Learn More

2-Quinolinol 98.0+%, TCI America™

Catalog No. H03041G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03041G 1 g
H030410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03041G Supplier TCI America Supplier No. H03041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-31-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00006743
InChI Key LISFMEBWQUVKPJ-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone
PubChem CID 6038
ChEBI CHEBI:18289
IUPAC Name 1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)N2

Specifications

Melting Point 197°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Quinolinol
RTECSNumber FG7175000
TSCA Yes