missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidone 98.0+%, TCI America™

Catalog No. P0575500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0575500G 500 g
P057525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0575500G Supplier TCI America Supplier No. P0575500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1

Specifications

Color White-Yellow
Boiling Point 245°C
Quantity 500 g
Formula Weight 85.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Pyrrolidone
EINECSNumber (5)-0112
RTECSNumber UY5715000
TSCA Yes