missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-ylpyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC69304DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC69304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

937796-10-6
177.21
KJMKROLYLRRJDN-UHFFFAOYSA-N
24229766
O=CC1=CN=C(N=C1)N1CCCC1
C9H11N3O
MFCD09863239
2-pyrrolidin-1-yl pyrimidine-5-carbaldehyde, 2-pyrrolidin-1-yl pyrimidine-5-carboxaldehyde, 2-1-pyrrolidinyl-5-pyrimidinecarbaldehyde, 2-pyrrolidinylpyrimidine-5-carbaldehyde, 2-pyrrolidin-1-ylpyrimidine-5-carboxaldehyde, 2-1-pyrrolidinyl-5-pyrimidinecarboxaldehyde, saltdata: free
2-pyrrolidin-1-ylpyrimidine-5-carbaldehyde

Specifications

2-Pyrrolidin-1-ylpyrimidine-5-carbaldehyde
96.5
97%
C9H11N3O
MFCD09863239
KJMKROLYLRRJDN-UHFFFAOYSA-N
2-pyrrolidin-1-ylpyrimidine-5-carbaldehyde
24229766
97%
937796-10-6
100.0
Amber Glass Bottle
1g
2-pyrrolidin-1-yl pyrimidine-5-carbaldehyde, 2-pyrrolidin-1-yl pyrimidine-5-carboxaldehyde, 2-1-pyrrolidinyl-5-pyrimidinecarbaldehyde, 2-pyrrolidinylpyrimidine-5-carbaldehyde, 2-pyrrolidin-1-ylpyrimidine-5-carboxaldehyde, 2-1-pyrrolidinyl-5-pyrimidinecarboxaldehyde, saltdata: free
O=CC1=CN=C(N=C1)N1CCCC1
177.21
177.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents