missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-ylisonicotinaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. CC50404DA
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC50404DA 1g Amber Glass Bottle
CC50404CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC50404DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC50404DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 898289-23-1
Formule moléculaire C10H12N2O
Poids moléculaire (g/mol) 176.219
Numéro MDL MFCD08690286
Clé InChI QPKFMHRNVDJMLZ-UHFFFAOYSA-N
Synonyme 2-pyrrolidin-1-ylisonicotinaldehyde, 2-pyrrolidin-1-yl isonicotinaldehyde, 2-pyrrolidin-1-yl pyridine-4-carbaldehyde, 2-pyrrolidin-1-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-1-pyrrolidinyl, 2-pyrrolidinylpyridine-4-carbaldehyde, 2-pyrrolidin-1-yl-isonicotinaldehyde, 4-pyridinecarboxaldehyde, 2-1-pyrrolidinyl
PubChem CID 18525845
Nom de l’IUPAC 2-pyrrolidin-1-ylpyridine-4-carbaldehyde
SOURIRES C1CCN(C1)C2=NC=CC(=C2)C=O

Spécifications

Nom ou substance chimique 2-Pyrrolidin-1-ylisonicotinaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 176.22
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.