missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinemethanol, 98+%

Catalog No. AAA1500536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500536 500 g
AAA1500514 25 g
AAA1500522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500536 Supplier Thermo Scientific Chemicals Supplier No. A1500536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 586-98-1 | C6H7NO | 109.128 g/mol

2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 586-98-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006348
InChI Key SHNUBALDGXWUJI-UHFFFAOYSA-N
Synonym 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol
PubChem CID 11474
IUPAC Name pyridin-2-ylmethanol
SMILES C1=CC=NC(=C1)CO

Specifications

Melting Point -6°C to -5°C
Density 1.131
Boiling Point 112°C to 113°C (16 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.544
Quantity 500 g
Beilstein 107849
Sensitivity Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 2-Pyridinemethanol
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 209-592-7
RTECSNumber UT4689000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only