missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3110300
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC36901CB 250mg Amber Glass Bottle
CC36901DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC36901CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC36901CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 116247-03-1
Formule moléculaire C9H8N2O2S
Poids moléculaire (g/mol) 208.24
Numéro MDL MFCD02854711
Clé InChI LJGAQGZEJDQDAU-UHFFFAOYNA-N
Synonyme 2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl, 2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid, 3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine, 2-3-pyridyl-1,3-thiazoline-4-carboxylic acid, acmc-20dw5p, 4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole
PubChem CID 3130734
Nom de l’IUPAC 2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
SOURIRES OC(=O)C1CSC(=N1)C1=CN=CC=C1

Spécifications

Nom ou substance chimique 2-Pyrid-3-yl-4, 5-dihydro-1, 3-thiazole-4-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 208.24
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.