missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, puriss., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of BP, Ph. Eur., USP,≥99.5% (GC)
$243.50 - $5178.60
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| MDL Number | MFCD00011674 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
60045530
|
Honeywell Chemicals
241371L |
1 L | Poly Bottle |
Each for $243.50
|
|
||||
|
60045531
|
Honeywell Chemicals
2413725L |
2.5 L | Poly Bottle |
Each for $350.85
|
|
||||
|
60045536
|
Honeywell Chemicals
241375L |
5 L | Poly Bottle |
Each for $616.85
|
|
||||
|
60045537
|
Honeywell Chemicals
241376X1L |
6 x 1 L | Poly Bottle |
Case of 6 Each for $1,083.13
|
|
||||
|
60045534
|
Honeywell Chemicals
241374X25L |
4 x 2.5 L | Poly Bottle |
Case of 4 Each for $1,154.35
|
|
||||
|
60045532
|
Honeywell Chemicals
2413720KG |
20 kg | Metal Drum |
Each for $2,274.35
|
|
||||
|
60045535
|
Honeywell Chemicals
241374X5L |
4 x 5 L | Poly Bottle |
Case of 4 Each for $2,139.00
|
|
||||
|
60045529
|
Honeywell Chemicals
24137160KG |
160 kg | Metal Drum |
Each for $5,178.60
|
|
||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00011674 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Specifications
| -85°C | |
| Colorless | |
| 82°C | |
| 0.02 | |
| 750°F | |
| Liquid | |
| (CH3)2CHOH | |
| 33mmHg (20°C), 44mmHg (25°C) | |
| 635639 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1g/mol | |
| puriss. |
| 67-63-0 | |
| 0.785g/mL at 25°C | |
| 1 L | |
| 12°C (53.6°F) | |
| Poly Bottle | |
| C3H8O | |
| MFCD00011674 | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% (GC) | |
| 2-Propanol, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
DOTInformation : class 3 - PG 2 - Isopropanol
EINECSNumber : 200-661-7
RTECSNumber : NT8050000