missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, puriss., ≥99.5% (GC), Honeywell Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -85°C |
| Color | Colorless |
| Density | 0.785g/mL at 25°C |
| Quantity | 5 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 33mmHg (20°C), 44mmHg (25°C) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P305 + P351 + P338 |
| Precautionary Statement | H225-H319-H336 |
| DOTInformation | class 3 - PG 2 - Isopropanol |
| EINECSNumber | 200-661-7 |
| RTECSNumber | NT8050000 |