missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, puriss., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of BP, Ph. Eur., USP,≥99.5% (GC)
Supplier: Honeywell Chemicals 24137160KG
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| 2-Propanol | |
| Meets analytical specification of Ph. Eur.BP, USP | |
| Colorless | |
| 82°C | |
| 0.02 | |
| 750°F | |
| Liquid | |
| (CH3)2CHOH | |
| 33mmHg (20°C), 44mmHg (25°C) | |
| 635639 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1g/mol | |
| puriss. |
| -85°C | |
| 67-63-0 | |
| 0.785g/mL at 25°C | |
| 160 kg | |
| 12°C (53.6°F) | |
| Metal Drum | |
| C3H8O | |
| MFCD00011674 | |
| UN1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% (GC) |
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00011674 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
DOTInformation : class 3 - PG 2 - Isopropanol
EINECSNumber : 200-661-7
RTECSNumber : NT8050000