missing translation for 'onlineSavingsMsg'
Learn More

2-Propanethiol 96.0+%, TCI America™

Catalog No. P0489500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0489500ML 500 mL
P048925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0489500ML Supplier TCI America Supplier No. P0489500ML

Chemical Identifiers

CAS 75-33-2
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004863
InChI Key KJRCEJOSASVSRA-UHFFFAOYSA-N
Synonym 2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
PubChem CID 6364
ChEBI CHEBI:8474
IUPAC Name propane-2-thiol
SMILES CC(C)S

Specifications

Melting Point -130°C
Color Colorless
Boiling Point 53°C
Quantity 500 mL
UN Number 2402
Formula Weight 76.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propanethiol
EINECSNumber (2)-0464
RTECSNumber TZ7302000
TSCA Yes