missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiophene 95.0+%, TCI America™

Catalog No. P13975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13975G 5 g
P13971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13975G Supplier TCI America Supplier No. P13975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 825-55-8
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.234
MDL Number MFCD00130080
InChI Key PJRGDKFLFAYRBV-UHFFFAOYSA-N
Synonym thiophene, 2-phenyl, 2-phenyl-thiophene, phenylthiophene, thiophene, phenyl, 2-thienylbenzene, 2-phenyithiophene, 2-phenylthiofuran, 2-phenylthiophene, acmc-209ppe
PubChem CID 69999
IUPAC Name 2-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=CS2

Specifications

Melting Point 36°C
Color White
Quantity 5 g
Formula Weight 160.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylthiophene
TSCA No
Recommended Storage Refrigerator