missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylquinoline, 99+%, Thermo Scientific™

Numéro de catalogue. AC297990050
Modifier l'affichage
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
AC297990050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC297990050 Fournisseur Thermo Scientific Chemicals Code fournisseur 297990050
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Nom ou substance chimique 2-Phenylquinoline
CAS 612-96-4
CAS % minimum 99.0
CAS % maximal 100.0
Point d’ébullition 363°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 99% min. (GC)
Emballage Glass bottle
Formule moléculaire C15H11N
Quantité 5g
Numéro MDL MFCD00011568
Synonyme quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940
Renseignements sur la solubilité Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,ethyl ether and acetone
Clé InChI FSEXLNMNADBYJU-UHFFFAOYSA-N
SOURIRES C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1
Nom de l’IUPAC 2-phenylquinoline
Poids moléculaire (g/mol) 205.26
PubChem CID 71545
Poids de formule 205.26
Pourcentage de pureté 99+%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 210-326-7

RUO – Research Use Only