missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66304DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC66304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

130161-46-5
184.198
AUTGLFSBJLERMV-UHFFFAOYSA-N
820135
C1=CC=C(C=C1)C2=NC=C(C=N2)C=O
C11H8N2O
MFCD03085924
2-phenyl-5-pyrimidinecarboxaldehye, 2-phenylpyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-phenyl, 2-phenyl-pyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde,2-phenyl, 2-phenyl-5-pyrimidinecarbaldehyde, 2-phenyl-5-pyrimidinecarboxaldehyde, acmc-1c230, 1-2-phenylpyrimidin-5-yl methanone
2-phenylpyrimidine-5-carbaldehyde

Specifications

2-Phenylpyrimidine-5-carbaldehyde
96.5
97%
C11H8N2O
MFCD03085924
AUTGLFSBJLERMV-UHFFFAOYSA-N
2-phenylpyrimidine-5-carbaldehyde
820135
97%
130161-46-5
100.0
Amber Glass Bottle
1g
2-phenyl-5-pyrimidinecarboxaldehye, 2-phenylpyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-phenyl, 2-phenyl-pyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde,2-phenyl, 2-phenyl-5-pyrimidinecarbaldehyde, 2-phenyl-5-pyrimidinecarboxaldehyde, acmc-1c230, 1-2-phenylpyrimidin-5-yl methanone
C1=CC=C(C=C1)C2=NC=C(C=N2)C=O
184.198
184.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents