missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyridine, 98%

Catalog No. AAL0786118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0786118 50 g
AAL0786109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0786118 Supplier Thermo Scientific Chemicals Supplier No. L0786118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1008-89-5 | C11H9N | 155.20 g/mol

2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylpyridine is used to produce 2-phenyl-pyridine-1-oxide. It is used in the manufacturing of organic light emitting diodes (OLEDs). It is used as an intermediate.

Solubility
Fully miscible with ethanol, acetone, toluene, methylene chloride. Not miscible in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 1008-89-5
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.20
MDL Number MFCD00006280,MFCD31699954
InChI Key VQGHOUODWALEFC-UHFFFAOYSA-N
Synonym pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
PubChem CID 13887
IUPAC Name 2-phenylpyridine
SMILES C1=CC=C(C=C1)C1=CC=CC=N1

Specifications

Melting Point -5°C
Density 1.086
Boiling Point 268°C to 270°C
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 50 g
Beilstein 110445
Solubility Information Fully miscible with ethanol,acetone,toluene,methylene chloride. Not miscible in water.
Formula Weight 155.2
Percent Purity 98%
Chemical Name or Material 2-Phenylpyridine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-763-1
RTECSNumber UT7140900
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only