missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindene, Thermo Scientific Chemicals

Catalog No. AC467770050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467770050 5 g
AC467770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467770050 Supplier Thermo Scientific Chemicals Supplier No. 467770050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4505-48-0
Molecular Formula C15H12
Molecular Weight (g/mol) 192.26
MDL Number MFCD00239514
InChI Key BSBXLZYWGGAVHD-UHFFFAOYSA-N
IUPAC Name 2-phenyl-1H-indene
SMILES C1C2=CC=CC=C2C=C1C1=CC=CC=C1

Specifications

Melting Point 157.0°C to 161.0°C
Quantity 5 g
Formula Weight 192.26
Chemical Name or Material 2-Phenylindene

RUO – Research Use Only