missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl Isobutyrate 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 250°C |
| Quantity | 25 mL |
| Formula Weight | 192.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Phenylethyl Isobutyrate |
Sécurité et manipulation
| EINECSNumber | (3)-1040&(3)-1045 |
| RTECSNumber | NQ5435000 |
| TSCA | Yes |