missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl Cinnamate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 57°C |
| Color | White-Yellow |
| Quantity | 500 g |
| Formula Weight | 252.31 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Phenylethyl Cinnamate |
Safety and Handling
| EINECSNumber | (3)-1739 |
| RTECSNumber | GE0405000 |
| TSCA | Yes |