missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylcarbamoyl-1,4-naphthoquinone-4-(4-diethylamino-2-methylphenyl)imine 98.0+%, TCI America™

Catalog No. P11495G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11495G Supplier TCI America Supplier No. P11495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102187-19-9
Molecular Formula C28H27N3O2
Molecular Weight (g/mol) 437.543
MDL Number MFCD00191571
InChI Key ZYKBEIDPRRYKKQ-UHFFFAOYSA-N
Synonym 3-Phenylcarbamoyl-N-[4′C-(N,N-diethylamino)-2′C-methylphenyl]-1,4-naphthoquinonimine, 4-[[4-(Diethylamino)-2-methylphenyl]imino]-1-oxo-N-phenyl-1,4-dihydronaphthalene-2-carboxamide
PubChem CID 634873
IUPAC Name 4-[4-(diethylamino)-2-methylphenyl]imino-1-oxo-N-phenylnaphthalene-2-carboxamide
SMILES CCN(CC)C1=CC(=C(C=C1)N=C2C=C(C(=O)C3=CC=CC=C32)C(=O)NC4=CC=CC=C4)C

Specifications

Melting Point 192°C
Color Green
Quantity 5 g
Formula Weight 437.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylcarbamoyl-1,4-naphthoquinone-4-(4-diethylamino-2-methylphenyl)imine
EINECSNumber (4)-0454
TSCA No