missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbutyrophenone 98.0+%, TCI America™

Catalog No. P22695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P22695G Supplier TCI America Supplier No. P22695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16282-16-9
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00010344
InChI Key UHKJKVIZTFFFSB-UHFFFAOYSA-N
Synonym 1,2-diphenyl-1-butanone, alpha-ethyldeoxybenzoin, alpha-ethyl deoxybenzoin, 2-phenylbutyrophenone, 1,2-diphenyl-butan-1-one, 1-butanone, 1,2-diphenyl, phenylbutyrophenone, pubchem20785, butyrophenone, 2-phenyl, 1-butanone,1,2-diphenyl
PubChem CID 297636
IUPAC Name 1,2-diphenylbutan-1-one
SMILES CCC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 56°C
Color White
Boiling Point 324°C
Quantity 5 g
Formula Weight 224.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylbutyrophenone