missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbutyric Acid 98.0+%, TCI America™

Catalog No. P0164500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0164500G 500 g
P016425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0164500G Supplier TCI America Supplier No. P0164500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-27-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002667
InChI Key OFJWFSNDPCAWDK-UHFFFAOYSA-N
Synonym 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl
PubChem CID 7012
ChEBI CHEBI:86545
IUPAC Name 2-phenylbutanoic acid
SMILES CCC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 43°C
Color White
Boiling Point 272°C
Quantity 500 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylbutyric Acid
EINECSNumber (3)-2951
RTECSNumber ET5957500
TSCA Yes